Assay Details
Gene Symbol: LOC101724434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870290.2 | XP_004870347.1 | 17 - 18 | 2349 | 68 | |
XM_013067629.2 | XP_012923083.1 | 24 - 25 | 3765 | 68 | ||
XM_021248594.1 | XP_021104253.1 | 23 - 24 | 3606 | 68 | ||
XM_021248596.1 | XP_021104255.1 | 24 - 25 | 3765 | 68 | ||
XM_021248597.1 | XP_021104256.1 | 24 - 25 | 3765 | 68 | ||
XM_021248598.1 | XP_021104257.1 | 22 - 23 | 3505 | 68 | ||
XM_021248599.1 | XP_021104258.1 | 24 - 25 | 3783 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
