Assay Details
Gene Symbol: LOC101724434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870290.2 | XP_004870347.1 | 19 - 20 | 2635 | 123 | |
XM_013067629.2 | XP_012923083.1 | 26 - 27 | 4051 | 123 | ||
XM_021248594.1 | XP_021104253.1 | 25 - 26 | 3892 | 123 | ||
XM_021248595.1 | XP_021104254.1 | 26 - 27 | 4048 | 123 | ||
XM_021248597.1 | XP_021104256.1 | 26 - 27 | 4051 | 123 | ||
XM_021248598.1 | XP_021104257.1 | 24 - 25 | 3791 | 123 | ||
XM_021248599.1 | XP_021104258.1 | 26 - 27 | 4069 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
