Assay Details
Gene Symbol: LOC101724434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870290.2 | XP_004870347.1 | 23 - 24 | 3043 | 71 | |
XM_013067629.2 | XP_012923083.1 | 30 - 31 | 4459 | 71 | ||
XM_021248594.1 | XP_021104253.1 | 29 - 30 | 4300 | 71 | ||
XM_021248595.1 | XP_021104254.1 | 30 - 31 | 4456 | 71 | ||
XM_021248596.1 | XP_021104255.1 | 29 - 30 | 4345 | 71 | ||
XM_021248597.1 | XP_021104256.1 | 30 - 31 | 4459 | 71 | ||
XM_021248598.1 | XP_021104257.1 | 28 - 29 | 4199 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
