Assay Details
Gene Symbol: Pigo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873924.3 | XP_004873981.1 | 6 - 7 | 2476 | 59 | |
XM_004873927.3 | XP_004873984.1 | 6 - 7 | 2447 | 59 | ||
XM_004873930.3 | XP_004873987.1 | 5 - 6 | 1936 | 59 | ||
XM_021251484.1 | XP_021107143.1 | 5 - 6 | 1514 | 59 | ||
XM_021251485.1 | XP_021107144.1 | 5 - 6 | 1512 | 59 | ||
XM_021251486.1 | XP_021107145.1 | 5 - 6 | 1521 | 59 | ||
XM_021251487.1 | XP_021107146.1 | 5 - 6 | 1519 | 59 | ||
XM_021251488.1 | XP_021107147.1 | 5 - 6 | 784 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
