Assay Details
| Gene Symbol: Hepacam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874482.3 | XP_004874539.1 | 3 - 4 | 845 | 67 | |
| XM_013069153.2 | XP_012924607.1 | 3 - 4 | 847 | 67 | ||
| XM_013069154.2 | XP_012924608.1 | 3 - 4 | 847 | 67 | ||
| XM_013069155.2 | XP_012924609.1 | 3 - 4 | 846 | 67 | ||
More Information
Set Membership: |
|

Hepacam
Genomic Map