Assay Details
Gene Symbol: Tmem131l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013067227.2 | XP_012922681.1 | 17 - 18 | 1867 | 53 | |
XM_013067228.2 | XP_012922682.1 | 17 - 18 | 1864 | 53 | ||
XM_013067229.2 | XP_012922683.1 | 17 - 18 | 1864 | 53 | ||
XM_021247869.1 | XP_021103528.1 | 17 - 18 | 1867 | 53 | ||
XM_021247871.1 | XP_021103530.1 | 17 - 18 | 1867 | 53 | ||
XM_021247872.1 | XP_021103531.1 | 17 - 18 | 1867 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
