Assay Details
| Gene Symbol: Npffr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021249821.1 | XP_021105480.1 | 2 - 3 | 600 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npffr2| Gene Symbol: Npffr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021249821.1 | XP_021105480.1 | 2 - 3 | 600 | 94 | |
Set Membership: |
|