Assay Details
| Gene Symbol: Agr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839594.3 | XP_004839651.1 | 7 - 8 | 625 | 63 | |
| XM_021254150.1 | XP_021109809.1 | 7 - 8 | 576 | 63 | ||
More Information
Set Membership: |
|

Agr2
Genomic Map