Assay Details
| Gene Symbol: LOC101720625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843060.3 | XP_004843117.1 | 12 - 13 | 1729 | 70 | |
| XM_004843061.2 | XP_004843118.1 | 11 - 12 | 1147 | 70 | ||
More Information
Set Membership: |
|

LOC101720625
Genomic Map