Assay Details
Gene Symbol: LOC101720625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843060.3 | XP_004843117.1 | 5 - 6 | 964 | 57 | |
XM_004843061.2 | XP_004843118.1 | 4 - 5 | 382 | 57 | ||
XM_013071573.2 | XP_012927027.1 | 5 - 6 | 964 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
