Assay Details
Gene Symbol: Adap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870591.3 | XP_004870648.2 | 4 - 5 | 456 | 58 | |
XM_021248901.1 | XP_021104560.1 | 4 - 5 | 456 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
