Assay Details
| Gene Symbol: LOC101720600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239518.1 | XP_021095177.1 | 4 - 5 | 573 | 97 | |
| XM_021239520.1 | XP_021095179.1 | 4 - 5 | 1256 | 97 | ||
| XM_021239521.1 | XP_021095180.1 | 4 - 5 | 581 | 97 | ||
| XM_021239522.1 | XP_021095181.1 | 3 - 4 | 798 | 97 | ||
| XM_021239523.1 | XP_021095182.1 | 3 - 4 | 483 | 97 | ||
| XM_021239524.1 | XP_021095183.1 | 3 - 4 | 545 | 97 | ||
| XM_021239525.1 | XP_021095184.1 | 3 - 4 | 493 | 97 | ||
| XM_021239526.1 | XP_021095185.1 | 4 - 5 | 382 | 97 | ||
| XM_021239527.1 | XP_021095186.1 | 4 - 5 | 392 | 97 | ||
| XM_021239528.1 | XP_021095187.1 | 4 - 5 | 444 | 97 | ||
| XM_021239529.1 | XP_021095188.1 | 2 - 3 | 509 | 97 | ||
| XM_021239530.1 | XP_021095189.1 | 3 - 4 | 322 | 97 | ||
| XM_021239534.1 | XP_021095193.1 | 4 - 5 | 583 | 97 | ||
More Information
Set Membership: |
|

LOC101720600
Genomic Map