Assay Details
Gene Symbol: LOC101721321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021242296.1 | XP_021097955.1 | 2 - 3 | 340 | 64 | |
XM_021242297.1 | XP_021097956.1 | 2 - 3 | 352 | 64 | ||
XM_021242298.1 | XP_021097957.1 | 2 - 3 | 326 | 64 |

More Information
Set Membership: |
![]() ![]() |
