Assay Details
Gene Symbol: LOC101721321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021242296.1 | XP_021097955.1 | 3 - 4 | 510 | 95 | |
XM_021242297.1 | XP_021097956.1 | 3 - 4 | 522 | 95 | ||
XM_021242298.1 | XP_021097957.1 | 3 - 4 | 496 | 95 | ||
XM_021242299.1 | XP_021097958.1 | 3 - 4 | 507 | 95 | ||
XM_021242301.1 | XP_021097960.1 | 3 - 4 | 519 | 95 | ||
XM_021242302.1 | XP_021097961.1 | 3 - 4 | 493 | 95 |
