Assay Details
Gene Symbol: LOC101721321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021242296.1 | XP_021097955.1 | 5 - 6 | 788 | 79 | |
XM_021242297.1 | XP_021097956.1 | 5 - 6 | 800 | 79 | ||
XM_021242298.1 | XP_021097957.1 | 5 - 6 | 774 | 79 | ||
XM_021242299.1 | XP_021097958.1 | 5 - 6 | 785 | 79 | ||
XM_021242301.1 | XP_021097960.1 | 5 - 6 | 797 | 79 | ||
XM_021242302.1 | XP_021097961.1 | 5 - 6 | 771 | 79 |
