Assay Details
Gene Symbol: Cbfa2t3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021256560.1 | XP_021112219.1 | 5 - 6 | 742 | 57 | |
XM_021256561.1 | XP_021112220.1 | 5 - 6 | 739 | 57 | ||
XM_021256562.1 | XP_021112221.1 | 4 - 5 | 664 | 57 | ||
XM_021256563.1 | XP_021112222.1 | 5 - 6 | 738 | 57 | ||
XM_021256564.1 | XP_021112223.1 | 5 - 6 | 647 | 57 | ||
XM_021256565.1 | XP_021112224.1 | 5 - 6 | 644 | 57 | ||
XM_021256566.1 | XP_021112225.1 | 5 - 6 | 741 | 57 | ||
XM_021256567.1 | XP_021112226.1 | 5 - 6 | 742 | 57 | ||
XM_021256568.1 | XP_021112227.1 | 4 - 5 | 664 | 57 | ||
XM_021256569.1 | XP_021112228.1 | 4 - 5 | 531 | 57 | ||
XM_021256570.1 | XP_021112229.1 | 4 - 5 | 544 | 57 | ||
XM_021256571.1 | XP_021112230.1 | 5 - 6 | 742 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
