Assay Details
Gene Symbol: LOC101704508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840531.3 | XP_004840588.1 | 2 - 3 | 771 | 60 | |
XM_021254818.1 | XP_021110477.1 | 2 - 3 | 753 | 60 |

More Information
Set Membership: |
![]() ![]() |
