Assay Details
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004841778.2 | XP_004841835.1 | 4 - 5 | 241 | 89 | |
XM_013071124.1 | XP_012926578.1 | 4 - 5 | 205 | 89 | ||
XM_013071125.2 | XP_012926579.1 | 4 - 5 | 211 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
