Assay Details
Gene Symbol: Bloc1s5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847718.3 | XP_004847775.1 | 4 - 5 | 464 | 94 | |
XM_004847719.3 | XP_004847776.1 | 4 - 5 | 445 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
