Assay Details
Gene Symbol: Cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004859579.2 | XP_004859636.1 | 7 - 8 | 798 | 63 | |
XM_021240077.1 | XP_021095736.1 | 7 - 8 | 801 | 63 | ||
XM_021240078.1 | XP_021095737.1 | 7 - 8 | 800 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
