Assay Details
Gene Symbol: LOC101703542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860824.3 | XP_004860881.1 | 1 - 2 | 562 | 77 | |
XM_021241142.1 | XP_021096801.1 | 1 - 2 | 565 | 77 | ||
XM_021241143.1 | XP_021096802.1 | 1 - 2 | 565 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
