Assay Details
| Gene Symbol: Slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865225.2 | XP_004865282.1 | 5 - 6 | 1450 | 61 | |
| XM_004865226.3 | XP_004865283.1 | 5 - 6 | 1363 | 61 | ||
More Information
Set Membership: |
|

Slc19a2
Genomic Map