Assay Details
Gene Symbol: Ttc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870781.3 | XP_004870838.1 | 6 - 7 | 539 | 73 | |
XM_004870782.3 | XP_004870839.1 | 6 - 7 | 592 | 73 | ||
XM_021248985.1 | XP_021104644.1 | 6 - 7 | 512 | 73 | ||
XM_021248986.1 | XP_021104645.1 | 5 - 6 | 1800 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
