Assay Details
Gene Symbol: LOC101706757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871188.3 | XP_004871245.2 | 2 - 3 | 312 | 80 | |
XM_021249537.1 | XP_021105196.1 | 2 - 3 | 317 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
