Assay Details
Gene Symbol: Nbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013077718.2 | XP_012933172.1 | 14 - 15 | 2122 | 55 | |
XM_013077719.2 | XP_012933173.1 | 14 - 15 | 1989 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
