Assay Details
Gene Symbol: Bend7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013078509.2 | XP_012933963.1 | 3 - 4 | 594 | 83 | |
XM_013078510.2 | XP_012933964.1 | 3 - 4 | 594 | 83 | ||
XM_013078511.2 | XP_012933965.1 | 3 - 4 | 595 | 83 | ||
XM_013078513.2 | XP_012933967.1 | 3 - 4 | 595 | 83 | ||
XM_013078514.2 | XP_012933968.1 | 3 - 4 | 560 | 83 | ||
XM_013078515.2 | XP_012933969.1 | 3 - 4 | 560 | 83 | ||
XM_013078516.2 | XP_012933970.1 | 3 - 4 | 972 | 83 | ||
XM_021242079.1 | XP_021097738.1 | 3 - 4 | 594 | 83 | ||
XM_021242080.1 | XP_021097739.1 | 3 - 4 | 516 | 83 | ||
XM_021242081.1 | XP_021097740.1 | 3 - 4 | 595 | 83 | ||
XM_021242082.1 | XP_021097741.1 | 3 - 4 | 597 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
