Assay Details
Gene Symbol: LOC101706346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021244592.1 | XP_021100251.1 | 6 - 7 | 977 | 70 | |
XM_021244593.1 | XP_021100252.1 | 4 - 5 | 1772 | 70 | ||
XM_021244594.1 | XP_021100253.1 | 7 - 8 | 858 | 70 | ||
XM_021244595.1 | XP_021100254.1 | 6 - 7 | 1534 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021244592.1, XM_021244593.1, XM_021244594.1, XM_021244595.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
