Assay Details
Gene Symbol: LOC101703188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021253061.1 | XP_021108720.1 | 12 - 13 | 1223 | 118 | |
XM_021253063.1 | XP_021108722.1 | 11 - 12 | 1069 | 118 | ||
XM_021253065.1 | XP_021108724.1 | 11 - 12 | 966 | 118 | ||
XM_021253066.1 | XP_021108725.1 | 12 - 13 | 1095 | 118 | ||
XM_021253067.1 | XP_021108726.1 | 12 - 13 | 934 | 118 | ||
XM_021253068.1 | XP_021108727.1 | 11 - 12 | 831 | 118 | ||
XM_021253069.1 | XP_021108728.1 | 11 - 12 | 815 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
