Assay Details
Gene Symbol: LOC101703188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021253061.1 | XP_021108720.1 | 13 - 14 | 1265 | 110 | |
XM_021253063.1 | XP_021108722.1 | 12 - 13 | 1111 | 110 | ||
XM_021253065.1 | XP_021108724.1 | 12 - 13 | 1008 | 110 | ||
XM_021253066.1 | XP_021108725.1 | 13 - 14 | 1137 | 110 | ||
XM_021253067.1 | XP_021108726.1 | 13 - 14 | 976 | 110 | ||
XM_021253068.1 | XP_021108727.1 | 12 - 13 | 873 | 110 | ||
XM_021253069.1 | XP_021108728.1 | 12 - 13 | 857 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
