Assay Details
Gene Symbol: LOC101703188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021253061.1 | XP_021108720.1 | 6 - 7 | 737 | 109 | |
XM_021253062.1 | XP_021108721.1 | 6 - 7 | 735 | 109 | ||
XM_021253063.1 | XP_021108722.1 | 5 - 6 | 583 | 109 | ||
XM_021253064.1 | XP_021108723.1 | 5 - 6 | 583 | 109 | ||
XM_021253065.1 | XP_021108724.1 | 5 - 6 | 480 | 109 | ||
XM_021253066.1 | XP_021108725.1 | 6 - 7 | 609 | 109 | ||
XM_021253067.1 | XP_021108726.1 | 6 - 7 | 448 | 109 | ||
XM_021253068.1 | XP_021108727.1 | 5 - 6 | 345 | 109 | ||
XM_021253069.1 | XP_021108728.1 | 5 - 6 | 329 | 109 | ||
XM_021253070.1 | XP_021108729.1 | 6 - 7 | 738 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
