Assay Details
| Gene Symbol: Ccdc122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844240.2 | XP_004844297.1 | 4 - 5 | 668 | 110 | |
| XM_021257892.1 | XP_021113551.1 | 4 - 5 | 896 | 110 | ||
| XM_021257893.1 | XP_021113552.1 | 4 - 5 | 635 | 110 | ||
| XM_021257894.1 | XP_021113553.1 | 4 - 5 | 662 | 110 | ||
| XM_021257895.1 | XP_021113554.1 | 3 - 4 | 550 | 110 | ||
| XM_021257896.1 | XP_021113555.1 | 2 - 3 | 465 | 110 | ||
More Information
Set Membership: |
|

Ccdc122
Genomic Map