Assay Details
| Gene Symbol: Mob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851616.3 | XP_004851673.1 | 4 - 5 | 545 | 66 | |
| XM_004851617.3 | XP_004851674.1 | 3 - 4 | 452 | 66 | ||
| XM_004851618.3 | XP_004851675.1 | 3 - 4 | 338 | 66 | ||
| XM_021263998.1 | XP_021119657.1 | 4 - 5 | 480 | 66 | ||
| XM_021263999.1 | XP_021119658.1 | 5 - 6 | 481 | 66 | ||
| XM_021264000.1 | XP_021119659.1 | 3 - 4 | 384 | 66 | ||
| XM_021264001.1 | XP_021119660.1 | 3 - 4 | 447 | 66 | ||
More Information
Set Membership: |
|

Mob2
Genomic Map