Assay Details
Gene Symbol: Znf516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004861935.3 | XP_004861992.1 | 2 - 3 | 2041 | 96 | |
XM_004861936.3 | XP_004861993.1 | 2 - 3 | 2022 | 96 | ||
XM_004861937.3 | XP_004861994.1 | 2 - 3 | 2045 | 96 | ||
XM_004861939.3 | XP_004861996.1 | 2 - 3 | 2041 | 96 | ||
XM_021242177.1 | XP_021097836.1 | 2 - 3 | 2601 | 96 | ||
XM_021242178.1 | XP_021097837.1 | 2 - 3 | 2009 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
