Assay Details
Gene Symbol: Znf516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004861935.3 | XP_004861992.1 | 4 - 5 | 3609 | 58 | |
XM_004861936.3 | XP_004861993.1 | 4 - 5 | 3590 | 58 | ||
XM_004861937.3 | XP_004861994.1 | 4 - 5 | 3613 | 58 | ||
XM_021242177.1 | XP_021097836.1 | 4 - 5 | 4169 | 58 | ||
XM_021242178.1 | XP_021097837.1 | 4 - 5 | 3577 | 58 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
