Assay Details
Gene Symbol: Tesk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870158.2 | XP_004870215.1 | 7 - 8 | 796 | 69 | |
XM_021248499.1 | XP_021104158.1 | 3 - 4 | 316 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
