Assay Details
Gene Symbol: LOC101713038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013070279.2 | XP_012925733.1 | 6 - 7 | 594 | 107 | |
XM_013070280.2 | XP_012925734.1 | 6 - 7 | 594 | 107 | ||
XM_021254096.1 | XP_021109755.1 | 6 - 7 | 564 | 107 | ||
XM_021254097.1 | XP_021109756.1 | 6 - 7 | 697 | 107 | ||
XM_021254098.1 | XP_021109757.1 | 6 - 7 | 657 | 107 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
