Assay Details
Gene Symbol: Cnnm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004844374.3 | XP_004844431.1 | 3 - 4 | 1751 | 69 | |
XM_021258023.1 | XP_021113682.1 | 3 - 4 | 1752 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
