Assay Details
| Gene Symbol: Maff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845618.3 | XP_004845675.1 | 1 - 2 | 282 | 66 | |
| XM_021258999.1 | XP_021114658.1 | 1 - 2 | 292 | 66 | ||
| XM_021259008.1 | XP_021114667.1 | 1 - 2 | 297 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004845618.3, XM_021258999.1, XM_021259008.1 transcripts.
Set Membership: |
|

Maff
Genomic Map