Assay Details
Gene Symbol: Hspa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846954.3 | XP_004847011.1 | 1 - 2 | 88 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004846954.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
