Assay Details
Gene Symbol: Mapk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855159.2 | XP_004855216.1 | 3 - 4 | 1802 | 54 | |
XM_004855160.3 | XP_004855217.1 | 3 - 4 | 1652 | 54 | ||
XM_004855162.3 | XP_004855219.1 | 3 - 4 | 1616 | 54 | ||
XM_013075972.2 | XP_012931426.1 | 4 - 5 | 1709 | 54 | ||
XM_013075973.2 | XP_012931427.1 | 4 - 5 | 1914 | 54 | ||
XM_021236626.1 | XP_021092285.1 | 3 - 4 | 1644 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
