Assay Details
Gene Symbol: Smyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857148.2 | XP_004857205.1 | 4 - 5 | 769 | 63 | |
XM_021238179.1 | XP_021093838.1 | 4 - 5 | 771 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
