Assay Details
Gene Symbol: LOC101710499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858012.3 | XP_004858069.1 | 5 - 6 | 611 | 95 | |
XM_013077164.2 | XP_012932618.1 | 5 - 6 | 611 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
