Assay Details
Gene Symbol: Sdr9c7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004861271.2 | XP_004861328.1 | 1 - 2 | 373 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
