Assay Details
Gene Symbol: Gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865243.2 | XP_004865300.1 | 4 - 5 | 853 | 64 | |
XM_013065767.2 | XP_012921221.1 | 4 - 5 | 838 | 64 | ||
XM_021244706.1 | XP_021100365.1 | 4 - 5 | 713 | 64 | ||
XM_021244707.1 | XP_021100366.1 | 3 - 4 | 425 | 64 | ||
XM_021244708.1 | XP_021100367.1 | 3 - 4 | 407 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
