Assay Details
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004869629.2 | XP_004869686.1 | 5 - 6 | 644 | 75 | |
XM_021248227.1 | XP_021103886.1 | 5 - 6 | 644 | 75 | ||
XM_021248228.1 | XP_021103887.1 | 5 - 6 | 488 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
