Assay Details
| Gene Symbol: Gtf2h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872842.3 | XP_004872899.1 | 2 - 3 | 175 | 84 | |
| XM_004872844.3 | XP_004872901.1 | 2 - 3 | 175 | 84 | ||
| XM_021250682.1 | XP_021106341.1 | 1 - 2 | 82 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021250682.1 transcripts.
Set Membership: |
|

Gtf2h2
Genomic Map