Assay Details
| Gene Symbol: LOC101703973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262256.1 | XP_021117915.1 | 6 - 7 | 585 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101703973| Gene Symbol: LOC101703973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262256.1 | XP_021117915.1 | 6 - 7 | 585 | 84 | |
Set Membership: |
|