Assay Details
| Gene Symbol: Slc22a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836616.2 | XP_004836673.1 | 5 - 6 | 1157 | 62 | |
| XM_004836617.3 | XP_004836674.1 | 5 - 6 | 1161 | 62 | ||
| XM_013079044.2 | XP_012934498.1 | 5 - 6 | 1156 | 62 | ||
More Information
Set Membership: |
|

Slc22a5
Genomic Map