Assay Details
| Gene Symbol: Glis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838229.3 | XP_004838286.1 | 6 - 7 | 2229 | 72 | |
| XM_004838230.3 | XP_004838287.1 | 6 - 7 | 1978 | 72 | ||
| XM_013069690.2 | XP_012925144.1 | 5 - 6 | 2067 | 72 | ||
| XM_013069691.2 | XP_012925145.1 | 5 - 6 | 1698 | 72 | ||
| XM_021252774.1 | XP_021108433.1 | 6 - 7 | 2229 | 72 | ||
| XM_021252776.1 | XP_021108435.1 | 7 - 8 | 1728 | 72 | ||
More Information
Set Membership: |
|

Glis3
Genomic Map