Assay Details
Gene Symbol: Trim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864950.2 | XP_004865007.1 | 3 - 4 | 589 | 66 | |
XM_021244544.1 | XP_021100203.1 | 3 - 4 | 589 | 66 | ||
XM_021244545.1 | XP_021100204.1 | 3 - 4 | 466 | 66 | ||
XM_021244546.1 | XP_021100205.1 | 3 - 4 | 524 | 66 | ||
XM_021244547.1 | XP_021100206.1 | 3 - 4 | 402 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
